N-Phenyl-o-phenylendiamin ≥98.0% (by GC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
N-(2-Aminophenyl)aniline, 2-Aminodiphenylamine, N1-Phenylbenzene-1,2-diamine
A2129-25G
A2129-5G
TCIAA2129-25GEA
846
DKK
TCIAA2129-25G
TCIAA2129-5G
N-Phenyl-o-phenylendiamin ≥98.0% (by GC, titration analysis)
N-Phenyl-o-phenylendiamin
Formel:
C₁₂H₁₂N₂ Kogepunkt: 108°C Smeltepunkt: 77…80 °C Massefylde: 1,234 g/cm³ (20 °C) |
MDL Number:
MFCD00007685 CAS-nummer: 534-85-0 EINECS: 208-606-9 |
|
Specifikationer Testresultater
Appearance | White to Light yellow to Light red powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 78.0 to 82.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...