1,4-Diacetylbenzen ≥99.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

D0066-25G D0066-5G
TCIAD0066-25GEA 1100 DKK
TCIAD0066-25G TCIAD0066-5G
1,4-Diacetylbenzen ≥99.0% (by GC)
1,4-Diacetylbenzen
Formel: C₁₀H₁₀O₂
Kogepunkt: 120°C
Smeltepunkt: 111…115 °C
MDL Number: MFCD00008750
CAS-nummer: 1009-61-6
EINECS: 213-769-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7b6ae2a266e4bfb7c244c3d8f6240f2be7b56da8&vid=7b6ae2a266e4bfb7c244c3d8f6240f2be7b56da8&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystaline
Purity (GC) min. 99.0 %
Melting point 113.0 to 115.0 deg-C
Solubility in hot Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR