1,3-Diacetoxybenzen ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: m-Phenylene di(acetate)

R0046-25G R0046-500G
TCIAR0046-25GEA 148 DKK
TCIAR0046-25G TCIAR0046-500G
1,3-Diacetoxybenzen ≥98.0% (by GC)
1,3-Diacetoxybenzen
Formel: C₁₀H₁₀O₄
Kogepunkt: 278°C
Smeltepunkt: 142…143 °C
Massefylde: 1,185 g/cm³ (20 °C)
MDL Number: MFCD00008701
CAS-nummer: 108-58-7
EINECS: 203-596-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5cf542d85952b58e6177eaa904b8d161b446276c&vid=5cf542d85952b58e6177eaa904b8d161b446276c&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR