1,3-Bis(dimethylamino)propan ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: N,N,N',N'-Tetramethyl-1,3-diaminopropane, TMPDA, N1,N1,N3,N3-Tetramethylpropane-1,3-diamine, 1,3-Bis(dimethylamino)propane, N,N,N',N'-Tetramethyltrimethylenediamine

T0548-25ML T0548-500ML T0548-100ML
TCIAT0548-25MLEA 132 DKK
TCIAT0548-25ML TCIAT0548-500ML TCIAT0548-100ML
1,3-Bis(dimethylamino)propan ≥98.0% (by GC, titration analysis)
1,3-Bis(dimethylamino)propan
Formel: C₇H₁₈N₂
Molekylvægt: 130.23 g/mol
Kogepunkt: 145°C
Flammepunkt: 56°C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00008337
CAS-nummer: 110-95-2
EINECS: 203-818-8
UN nummer: 2734
ADR: 8 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ea10b11efe00e77c0b6a9dcd0845f5d0e557a622&vid=ea10b11efe00e77c0b6a9dcd0845f5d0e557a622&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR