1,3-Bis(dimethylamino)butan ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: N,N,N',N'-Tetramethyl-1,3-diaminobutane, N,N,N',N',1-Pentamethyltrimethylenediamine, N,N,N',N'-Tetramethyl-1,3-butanediamine

T0524-25ML T0524-500ML
TCIAT0524-25MLEA 411 DKK
TCIAT0524-25ML TCIAT0524-500ML
1,3-Bis(dimethylamino)butan ≥98.0% (by GC)
1,3-Bis(dimethylamino)butan
Formel: C₈H₂₀N₂
Molekylvægt: 144.26 g/mol
Kogepunkt: 169°C
Flammepunkt: 41°C
MDL Number: MFCD00025678
CAS-nummer: 97-84-7
EINECS: 202-610-4
UN nummer: 2733
ADR: 3 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0f1dd34983ad0dedb6145d24cb144fc74d23ec8d&vid=0f1dd34983ad0dedb6145d24cb144fc74d23ec8d&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR