4-Nitropyrocatechol ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 4-Nitrocatechol, 4-Nitrobenzene-1,2-diol

N0273-25G N0273-5G
TCIAN0273-25GEA 1070 DKK
TCIAN0273-25G TCIAN0273-5G
4-Nitropyrocatechol ≥98.0% (by GC, titration analysis)
4-Nitropyrocatechol
Formel: C₆H₅NO₄
MDL Number: MFCD00007242
CAS-nummer: 3316-09-4
EINECS: 222-009-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b388c0f0664daa5d53e12e451ed61162bc19e5de&vid=b388c0f0664daa5d53e12e451ed61162bc19e5de&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Light yellow to Amber to Dark green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 172.0 to 177.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR