(3,4-Dimethoxyphenyl)acetonitril ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 3,4-Dimethoxyphenylacetonitrile, Homoveratronitrile, 3,4-Dimethoxybenzyl cyanide

D1887-25G D1887-500G
TCIAD1887-25GEA 148 DKK
TCIAD1887-25G TCIAD1887-500G
(3,4-Dimethoxyphenyl)acetonitril ≥98.0% (by GC)
(3,4-Dimethoxyphenyl)acetonitril
Formel: C₁₀H₁₁NO₂
Kogepunkt: 160°C
Smeltepunkt: 60…62 °C
MDL Number: MFCD00001911
CAS-nummer: 93-17-4
EINECS: 202-225-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6e00449a0631ae806130b7238cb0412479bf470c&vid=6e00449a0631ae806130b7238cb0412479bf470c&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 62.0 to 66.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR