α-Brom-o-toluonitril ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 2-Cyanobenzyl bromide, 2-(Bromomethyl)benzonitrile, o-(Bromomethyl)benzonitrile

C1556-10G C1556-25G
TCIAC1556-10GEA 444 DKK
TCIAC1556-10G TCIAC1556-25G
α-Brom-o-toluonitril ≥98.0% (by GC)
α-Brom-o-toluonitril
Formel: C₈H₆BrN
Kogepunkt: 111°C
Smeltepunkt: 72…74 °C
MDL Number: MFCD00001794
CAS-nummer: 22115-41-9
UN nummer: 3261
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a0e87c9cd8bb953159b061b115204431da0df0c1&vid=a0e87c9cd8bb953159b061b115204431da0df0c1&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 72.0 to 74.0 deg-C
Solubility in Methanol very faint turbidity

Specifikationer

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR