N-(brommethyl)phthalimid ≥96.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 2-(Bromomethyl)isoindoline-1,3-dione

P1192-25G P1192-5G
TCIAP1192-25GEA 559 DKK
TCIAP1192-25G TCIAP1192-5G
N-(brommethyl)phthalimid ≥96.0% (by titrimetric analysis)
N-(brommethyl)phthalimid
Formel: C₉H₆BrNO₂
Molekylvægt: 240.06 g/mol
Kogepunkt: 410
MDL Number: MFCD00005897
CAS-nummer: 5332-26-3
EINECS: 226-239-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=91e46f64a6d92595e9d5f63830e7ba15e7d233c7&vid=91e46f64a6d92595e9d5f63830e7ba15e7d233c7&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 93.0 %
Purity (Argentmetric Titration) min. 96.0 %
Melting point 147.0 to 153.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR