(Benzyl)triphenylphosphonium chlorid ≥98.0% (by HPLC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
Benzyltriphenylphosphonium chloride
B0824-25G
B0824-500G
TCIAB0824-25GEA
312
DKK
TCIAB0824-25G
TCIAB0824-500G
(Benzyl)triphenylphosphonium chlorid ≥98.0% (by HPLC, titration analysis)
(Benzyl)triphenylphosphonium chlorid
Formel:
C₂₅H₂₂ClP Molekylvægt: 388.88 g/mol |
MDL Number:
MFCD00011913 CAS-nummer: 1100-88-5 EINECS: 214-154-3 UN nummer: 3464 ADR: 6.1 II |
|
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Argentmetric Titration) | min. 98.0 % |
Solubility in Water | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...