p-Tolyleddikesyre ≥98.0% (by GC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
2-(p-Tolyl)acetic acid, 4-Tolylacetic acid, (4-Methylphenyl)acetic acid
T0882-25G
T0882-250G
TCIAT0882-25GEA
189
DKK
TCIAT0882-25G
TCIAT0882-250G
p-Tolyleddikesyre ≥98.0% (by GC, titration analysis)
p-Tolyleddikesyre
Formel:
(C₆H₄(CH₃))CH₂COOH Molekylvægt: 150.18 g/mol Kogepunkt: 267°C Smeltepunkt: 91 |
MDL Number:
MFCD00004353 CAS-nummer: 622-47-9 EINECS: 210-738-7 |
|
Specifikationer Testresultater
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 92.0 to 94.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...