N-Isopropylbenzylamin ≥98.0% (by GC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
N-Benzyl-N-isopropylamine, N-Benzylisopropylamine
B1216-25ML
B1216-500ML
TCIAB1216-25MLEA
189
DKK
TCIAB1216-25ML
TCIAB1216-500ML
N-Isopropylbenzylamin ≥98.0% (by GC, titration analysis)
N-Isopropylbenzylamin
Formel:
C₆H₅CH₂N(CH₃)(CH₂CH₃) Molekylvægt: 149.24 g/mol Kogepunkt: 103°C Smeltepunkt: 0 Flammepunkt: 72°C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00008863 CAS-nummer: 102-97-6 EINECS: 203-067-6 |
|
Specifikationer Testresultater
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...