D(-)-Serin ≥98.5% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

S0033-25G S0033-5G
TCIAS0033-25GEA 411 DKK
TCIAS0033-25G TCIAS0033-5G
D(-)-Serin ≥98.5% (by titrimetric analysis)
D(-)-Serin
Formel: C₃H₇NO₃
Smeltepunkt: 235 °C
MDL Number: MFCD00004269
CAS-nummer: 312-84-5
EINECS: 206-229-4
Merck Index: 12,08604
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dad51a80202c1a334c9909fe58b339861731a9d1&vid=dad51a80202c1a334c9909fe58b339861731a9d1&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.5 %
Specific rotation [a]20/D -14.3 to -15.5 deg (C=10, HCl (1+5))
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR