(2-Fluorphenyl)(hydroxy)eddikkesyre ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

F0730-25G F0730-5G
TCIAF0730-25GEA 1690 DKK
TCIAF0730-25G TCIAF0730-5G
(2-Fluorphenyl)(hydroxy)eddikkesyre ≥98.0% (by GC, titration analysis)
(2-Fluorphenyl)(hydroxy)eddikkesyre
Formel: C₈H₇FO₃
Kogepunkt: 72°C
MDL Number: MFCD03095382
CAS-nummer: 389-31-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=48aed28a9a6d52ca51fca0a1a1923458a1453c7b&vid=48aed28a9a6d52ca51fca0a1a1923458a1453c7b&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 115.0 to 119.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR