2-Chlor-4-nitropyridin ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

C2283-1G C2283-25G C2283-5G
TCIAC2283-1GEA 362 DKK
TCIAC2283-1G TCIAC2283-25G TCIAC2283-5G
2-Chlor-4-nitropyridin ≥98.0% (by GC)
2-Chloro-4-nitropyridine
Formel: C₅H₃ClN₂O₂
MDL Number: MFCD00661454
CAS-nummer: 23056-36-2
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=890be5df1425423f2f0644ef14b170855128cfb3&vid=890be5df1425423f2f0644ef14b170855128cfb3&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 52.0 to 56.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR