1-Acetylnaphthalen ≥95.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 1'-Acetonaphthone, α-acetonaphthone, 1'-Acetylnaphthalene, Methyl α-naphthyl ketone, Methyl 1-naphthyl ketone, 1-Acetonaphthone

A1044-25G A1044-500G
TCIAA1044-25GEA 165 DKK
TCIAA1044-25G TCIAA1044-500G
1-Acetylnaphthalen ≥95.0% (by GC)
1-Acetylnaphthalen
Formel: C₁₂H₁₀O
Kogepunkt: 298°C
Smeltepunkt: 9…11 °C
Massefylde: 1,12 g/cm³ (20 °C)
Flammepunkt: ≥ 100 °C
MDL Number: MFCD00004013
CAS-nummer: 941-98-0
EINECS: 213-384-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7b3a625e5dc0997debceba4a19819fc0ceb8caf2&vid=7b3a625e5dc0997debceba4a19819fc0ceb8caf2&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Yellow to Orange clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR