Diisopropylsulfid ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Isopropyl sulphide

I0175-10ML I0175-25ML
TCIAI0175-10MLEA 197 DKK
TCIAI0175-10ML TCIAI0175-25ML
Diisopropylsulfid ≥98.0% (by GC)
Diisopropylsulfid
Formel: C₆H₁₄S
Molekylvægt: 118.24 g/mol
Kogepunkt: 121°C
Smeltepunkt: -69.1
Flammepunkt: 7°C
MDL Number: MFCD00008895
CAS-nummer: 625-80-9
EINECS: 210-911-7
UN nummer: 1993
ADR: 3 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=54d755e8c1f095077ad9de96af9e6de869d2d325&vid=54d755e8c1f095077ad9de96af9e6de869d2d325&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR