Triphenylphosphinsulfid ≥99.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

T1066-25G T1066-500G T1066-100G
TCIAT1066-25GEA 296 DKK
TCIAT1066-25G TCIAT1066-500G TCIAT1066-100G
Triphenylphosphinsulfid ≥99.0% (by GC)
Triphenylphosphine sulphide
Formel: C₁₈H₁₅PS
Molekylvægt: 294.36 g/mol
MDL Number: MFCD00004917
CAS-nummer: 3878-45-3
EINECS: 223-407-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b8bdef6ab14952eb2724efe10ac6eecc71527544&vid=b8bdef6ab14952eb2724efe10ac6eecc71527544&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 99.0 %
Melting point 162.0 to 165.0 deg-C
Solubility in hot Toluene almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR