Tetrabutylammoniumperchlorat ≥98.0% (by titrimetric analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
Tetra-n-butylammonium perchlorate
T0836-25G
T0836-500G
T0836-100G
TCIAT0836-25GEA
386
DKK
TCIAT0836-25G
TCIAT0836-500G
TCIAT0836-100G
Tetrabutylammoniumperchlorat ≥98.0% (by titrimetric analysis)
Tetrabutylammonium perchlorate
Formel:
C₁₆H₃₆ClNO₄ Molekylvægt: 341.92 g/mol Smeltepunkt: 210 |
MDL Number:
MFCD00038722 CAS-nummer: 1923-70-2 EINECS: 217-655-5 UN nummer: 1479 ADR: 5.1 II |
|
Specifikationer Testresultater
Appearance | White to Almost white powder to crystaline |
Purity (Ion exchange titration) | min. 98.0 % |
Purity (with Total Nitrogen) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...