Cyclopropancarbonitril ≥97.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Cyclopropyl cyanide

C0997-25ML C0997-5ML
TCIAC0997-25MLEA 468 DKK
TCIAC0997-25ML TCIAC0997-5ML
Cyclopropancarbonitril ≥97.0% (by GC)
Cyclopropancarbonitril
Formel: C₄H₅N
Kogepunkt: 135°C
Smeltepunkt: –25 °C
Massefylde: 0,911 g/cm³ (20 °C)
Flammepunkt: 40°C
MDL Number: MFCD00001269
CAS-nummer: 5500-21-0
EINECS: 226-836-8
UN nummer: 3275
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f757ad091c1a28684103735b9fe684d31377fceb&vid=f757ad091c1a28684103735b9fe684d31377fceb&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Red to Green clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR