Tetrafluor-p-benzoquinon ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Tetrafluoroquinone, 2,3,5,6-Tetrafluoro-1,4-benzoquinone, Tetrafluoro-1,4-benzoquinone, p-Fluoranil, 4-Fluoranil

T0790-1G T0790-5G
TCIAT0790-1GEA 321 DKK
TCIAT0790-1G TCIAT0790-5G
Tetrafluor-p-benzoquinon ≥98.0% (by HPLC, titration analysis)
Tetrafluor-p-benzoquinon
Formel: C₆F₄O₂
Molekylvægt: 180.06 g/mol
Kogepunkt: 133
MDL Number: MFCD00001592
CAS-nummer: 527-21-9
EINECS: 208-411-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2afd389cb437f9c72827b973bcc583edb2d05bf0&vid=2afd389cb437f9c72827b973bcc583edb2d05bf0&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Light yellow to Brown powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Iodometric Titration) min. 98.0 %
Solubility in Toluene almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR