6-tert-Butyl-o-cresol ≥99.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 2-(tert-Butyl)-6-methylphenol, 2-tert-Butyl-6-methylphenol

B0728-25G B0728-500G
TCIAB0728-25GEA 132 DKK
TCIAB0728-25G TCIAB0728-500G
6-tert-Butyl-o-cresol ≥99.0% (by GC)
6-tert-Butyl-o-cresol
Formel: C₁₁H₁₆O
Molekylvægt: 164.25
Kogepunkt: 227
Massefylde: 0.967
MDL Number: MFCD00002239
CAS-nummer: 2219-82-1
EINECS: 218-734-7
UN nummer: 2430
ADR: 8 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=63a8e3aca2917e7627c6b7f29cd70543ade20691&vid=63a8e3aca2917e7627c6b7f29cd70543ade20691&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White or Colorles to Yellow to Orange powder to lump to clear liquid
Purity (GC) min. 99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR