1,1-Dimethoxy-2-phenylethan ≥97.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
Phenylacetaldehyde dimethyl acetal
P1161-25ML
P1161-500ML
TCIAP1161-25MLEA
132
DKK
TCIAP1161-25ML
TCIAP1161-500ML
1,1-Dimethoxy-2-phenylethan ≥97.0% (by GC)
1,1-Dimethoxy-2-phenylethan
Formel:
C₆H₅CH₂CH(OCH₃)₂ Kogepunkt: 108°C Smeltepunkt: 62…64 °C Massefylde: 1 g/cm³ (20 °C) Flammepunkt: 90°C |
MDL Number:
MFCD00008487 CAS-nummer: 101-48-4 EINECS: 202-945-6 |
|
Specifikationer Testresultater
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...