5'-Fluor-2'-hydroxyacetophenon ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 1-(5-Fluoro-2-hydroxyphenyl)ethanone

F0318-25G F0318-5G
TCIAF0318-25GEA 665 DKK
TCIAF0318-25G TCIAF0318-5G
5'-Fluor-2'-hydroxyacetophenon ≥98.0% (by GC)
5'-Fluor-2'-hydroxyacetophenon
Formel: C₈H₇FO₂
Molekylvægt: 154.14 g/mol
Kogepunkt: 100°C
Smeltepunkt: 56
MDL Number: MFCD00011668
CAS-nummer: 394-32-1
EINECS: 206-893-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6942d3f2fa13fda7eed9d34bc426cc854cef4e95&vid=6942d3f2fa13fda7eed9d34bc426cc854cef4e95&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Gray to Brown powder to crystal
Purity (GC) min. 98.0 %
Melting point 56.0 to 60.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR