4'-Hydroxy-3'-methoxyacetophenon ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

H0261-25G H0261-100G
TCIAH0261-25GEA 394 DKK
TCIAH0261-25G TCIAH0261-100G
4'-Hydroxy-3'-methoxyacetophenon ≥98.0% (by GC)
4'-Hydroxy-3'-methoxyacetophenon
Formel: C₉H₁₀O₃
Kogepunkt: 300°C
Smeltepunkt: 115 °C
MDL Number: MFCD00008747
CAS-nummer: 498-02-2
EINECS: 207-854-5
Merck Index: 14,00741
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=03b486da5a9d3b23478de4470c73dee7f81015e0&vid=03b486da5a9d3b23478de4470c73dee7f81015e0&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 113.0 to 117.0 deg-C
Solubility in Methanol faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR