ω-Chlor-4-Fluoracetophenon ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
2-Chloro-1-(4-fluorophenyl)ethan-1-one, α-Chloro-4-fluoroacetophenone, 2-Chloro-1-(4-fluorophenyl)ethanone, Chloromethyl p-fluorophenyl ketone, 4-Fluorophenacyl chloride, 2-Chloro-4'-fluoroacetophenone
C1977-25G
C1977-5G
TCIAC1977-25GEA
411
DKK
TCIAC1977-25G
TCIAC1977-5G
ω-Chlor-4-Fluoracetophenon ≥98.0% (by GC)
ω-Chlor-4-Fluoracetophenon
Formel:
C₈H₆ClFO Molekylvægt: 172.59 g/mol Kogepunkt: 247 Smeltepunkt: 45 |
MDL Number:
MFCD00011652 CAS-nummer: 456-04-2 EINECS: 207-256-4 |
|
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 48.0 to 52.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...