2,6-Dibrom-p-toluidin ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 4-Amino-3,5-dibromotoluene, 2,6-Dibromo-4-methylaniline

D3577-25G D3577-5G
TCIAD3577-25GEA 1030 DKK
TCIAD3577-25G TCIAD3577-5G
2,6-Dibrom-p-toluidin ≥98.0% (by GC)
2,6-Dibrom-p-toluidin
Formel: C₇H₇Br₂N
Kogepunkt: 296 °C (1013 hPa)
Smeltepunkt: 79 °C
MDL Number: MFCD00007641
CAS-nummer: 6968-24-7
EINECS: 230-182-9

Bestil nu

Specifikationer Testresultater

Appearance White to Light red to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 72.0 to 76.0 deg-C

Specifikationer

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR