2-Nitro-p-anisidin ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 4-Amino-3-nitroanisole, 4-Methoxy-2-nitroaniline

M0119-25G M0119-500G M0119-100G
TCIAM0119-25GEA 173 DKK
TCIAM0119-25G TCIAM0119-500G TCIAM0119-100G
2-Nitro-p-anisidin ≥98.0% (by GC)
2-Nitro-p-anisidin
Formel: C₇H₈N₂O₃
Molekylvægt: 168.15 g/mol
MDL Number: MFCD00007152
CAS-nummer: 96-96-8
EINECS: 202-547-2
UN nummer: 2811
ADR: 6.1 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c65ea93b4625e221c55fc66ffd8ddeb924c77048&vid=c65ea93b4625e221c55fc66ffd8ddeb924c77048&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Orange to Red powder to crystal
Purity (GC) min. 98.0 %
Melting point 123.0 to 127.0 deg-C
Solubility in hot Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR