4-Fluor-2-iodanilin ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

F0706-25G F0706-5G
TCIAF0706-25GEA 1170 DKK
TCIAF0706-25G TCIAF0706-5G
4-Fluor-2-iodanilin ≥98.0% (by GC, titration analysis)
4-Fluor-2-iodanilin
Formel: C₆H₅FIN
Molekylvægt: 237.02 g/mol
MDL Number: MFCD00042584
CAS-nummer: 61272-76-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=21f64be812ce69302624ce7dcd45d4d5087ef50b&vid=21f64be812ce69302624ce7dcd45d4d5087ef50b&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Brown clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR