2-Fluor-4-iodanilin ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

F0417-25G F0417-5G
TCIAF0417-25GEA 485 DKK
TCIAF0417-25G TCIAF0417-5G
2-Fluor-4-iodanilin ≥98.0% (by GC)
2-Fluor-4-iodanilin
Formel: C₆H₅FIN
Molekylvægt: 237.02 g/mol
Kogepunkt: 259
MDL Number: MFCD00011738
CAS-nummer: 29632-74-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=18c6d4cb70029784381d1997881b8a21aff4f23d&vid=18c6d4cb70029784381d1997881b8a21aff4f23d&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Gray to Brown powder to crystal
Purity (GC) min. 98.0 %
Melting point 52.0 to 56.0 deg-C

Specifikationer

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR