Octyl-3,4,5-trihydroxybenzoat ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Octyl 3,4,5-trihydroxybenzoate, Gallic acid octyl ester, Octyl gallate, n-Octyl gallate

G0206-25G G0206-250G
TCIAG0206-25GEA 271 DKK
TCIAG0206-25G TCIAG0206-250G
Octyl-3,4,5-trihydroxybenzoat ≥98.0% (by titrimetric analysis)
Octyl-3,4,5-trihydroxybenzoat
Formel: C₁₅H₂₂O₅
Molekylvægt: 282.34 g/mol
MDL Number: MFCD00002197
CAS-nummer: 1034-01-1
EINECS: 213-853-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b36c29337f140113485a6207c407478a7b63c1da&vid=b36c29337f140113485a6207c407478a7b63c1da&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow to Light orange powder to crystaline
Purity (Neutralization titration) min. 98.0 %
Melting point 95.0 to 104.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR