Ethyl 3,4,5-trihydroxybenzoat ≥98.0% (by titrimetric analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
Gallic acid ethyl ester, Ethyl 3,4,5-trihydroxybenzoate, Ethyl gallate
G0016-25G
G0016-500G
G0016-100G
TCIAG0016-25GEA
189
DKK
TCIAG0016-25G
TCIAG0016-500G
TCIAG0016-100G
Ethyl 3,4,5-trihydroxybenzoat ≥98.0% (by titrimetric analysis)
Ethyl 3,4,5-trihydroxybenzoat
Formel:
C₉H₁₀O₅ Molekylvægt: 198.18 g/mol |
MDL Number:
MFCD00016430 CAS-nummer: 831-61-8 EINECS: 212-608-5 |
|
Specifikationer Testresultater
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (Neutralization titration) | 98.0 to 101.0 % |
Melting point | 149.0 to 154.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...