2,6-Dichlor-4-Fluoranilin ≥97.0% (by GC, total nitrogen)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

D3625-1G D3625-5G
TCIAD3625-1GEA 214 DKK
TCIAD3625-1G TCIAD3625-5G
2,6-Dichlor-4-Fluoranilin ≥97.0% (by GC, total nitrogen)
2,6-Dichlor-4-Fluoranilin
Formel: C₆H₄Cl₂FN
Molekylvægt: 180.01 g/mol
MDL Number: MFCD00142845
CAS-nummer: 344-19-4
UN nummer: 2811
ADR: 6.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a1aca3d0c47b71595a1359de7e14020790d0acbe&vid=a1aca3d0c47b71595a1359de7e14020790d0acbe&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Brown powder to crystal
Purity (GC) min. 97.0 %
Purity (with Total Nitrogen) min. 97.0 %
Melting point 54.0 to 58.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR