4-Brom-2-nitroanilin ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
B3033-25G
B3033-5G
TCIAB3033-25GEA
682
DKK
TCIAB3033-25G
TCIAB3033-5G
4-Brom-2-nitroanilin ≥98.0% (by GC)
4-Brom-2-nitroanilin
Formel:
C₆H₅BrN₂O₂ Kogepunkt: 309 °C (1013 hPa) Smeltepunkt: 110…113 °C Massefylde: 1,81 g/cm³ (20 °C) |
MDL Number:
MFCD00041312 CAS-nummer: 875-51-4 |
|
Specifikationer Testresultater
Appearance | Light yellow to Brown powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 110.0 to 114.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...