2-Phenylethylacetat ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Phenylethyl Acetate, 2-Phenethyl acetate, Phenethyl acetate

A0692-25G A0692-500G
TCIAA0692-25GEA 132 DKK
TCIAA0692-25G TCIAA0692-500G
2-Phenylethylacetat ≥98.0% (by GC)
2-Phenylethylacetat
Formel: C₁₀H₁₂O₂
Kogepunkt: 233 °C (1013 hPa)
Smeltepunkt: –31 °C
Massefylde: 1,0286 g/cm³ (20 °C)
Flammepunkt: 110°C
MDL Number: MFCD00008720
CAS-nummer: 103-45-7
EINECS: 203-113-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dec7b270a57ee4a9ec1755eca69d414bd0259de3&vid=dec7b270a57ee4a9ec1755eca69d414bd0259de3&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR