1-Pentansulfonsyre natriumsalt ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Sodium 1-pentanesulphonate, Sodium pentane-1-sulphonate, Pentane-1-sulphonic acid sodium salt, Sodium pentanesulphonate, 98%, Sodium pentanesulphonate

P0343-25G P0343-500G P0343-100G
TCIAP0343-25GEA 534 DKK
TCIAP0343-25G TCIAP0343-500G TCIAP0343-100G
1-Pentansulfonsyre natriumsalt ≥98.0% (by titrimetric analysis)
1-Pentansulfonsyre natriumsalt
Formel: C₅H₁₁O₃SNa
Smeltepunkt: > 300 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00007541
CAS-nummer: 22767-49-3
EINECS: 245-208-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f47522a22a7bb826167f3398da9ef648a2d36a70&vid=f47522a22a7bb826167f3398da9ef648a2d36a70&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White powder to crystal
Purity (Ion exchange titration) min. 98.0 %
Solubility in Water almost transparency
Drying loss max. 1.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR