Dimethylacetal ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
Acetaldehyddimethylacetal, 1,1-Dimethoxyethan
D0633-25ML
D0633-500ML
TCIAD0633-25MLEA
296
DKK
TCIAD0633-25ML
TCIAD0633-500ML
Dimethylacetal ≥98.0% (by GC)
Dimethylacetal
Formel:
C₄H₁₀O₂ Molekylvægt: 90.12 g/mol Kogepunkt: 65°C Smeltepunkt: –113°C Flammepunkt: 1°C |
MDL Number:
MFCD00008493 CAS-nummer: 534-15-6 EINECS: 208-589-8 UN nummer: 2377 ADR: 3 II Merck Index: 14,03226 |
|
Specifikationer Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...