Propylbutyrat ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
Butyric acid Propyl ester
B0764-25ML
B0764-500ML
TCIAB0764-25MLEA
132
DKK
TCIAB0764-25ML
TCIAB0764-500ML
Propylbutyrat ≥98.0% (by GC)
Propylbutyrat
Formel:
C₇H₁₄O₂ Molekylvægt: 130.19 g/mol Kogepunkt: 144°C Flammepunkt: 37°C |
MDL Number:
MFCD00009396 CAS-nummer: 105-66-8 EINECS: 203-320-0 UN nummer: 3272 ADR: 3 III Merck Index: 14,07846 |
|
Specifikationer Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...