Hexylbutyrat ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Butyric acid hexyl ester

B1891-25ML B1891-500ML
TCIAB1891-25MLEA 132 DKK
TCIAB1891-25ML TCIAB1891-500ML
Hexylbutyrat ≥98.0% (by GC)
Hexylbutyrat
Formel: C₁₀H₂₀O₂
Molekylvægt: 172.27 g/mol
Kogepunkt: 205°C
Flammepunkt: 81°C
MDL Number: MFCD00048884
CAS-nummer: 2639-63-6
EINECS: 220-136-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e157b59254daf73560e1e2ed0eb75eb252d2ac43&vid=e157b59254daf73560e1e2ed0eb75eb252d2ac43&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR