Isobutylbutyrat ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Butyric acid Isobutyl ester, 2-Methyl-1-propylbutyrate

B0761-25ML B0761-500ML
TCIAB0761-25MLEA 132 DKK
TCIAB0761-25ML TCIAB0761-500ML
Isobutylbutyrat ≥98.0% (by GC)
Isobutylbutyrat
Formel: C₈H₁₆O₂
Molekylvægt: 144.21 g/mol
Kogepunkt: 157°C
Flammepunkt: 49°C
MDL Number: MFCD00027134
CAS-nummer: 539-90-2
EINECS: 208-729-8
UN nummer: 3272
ADR: 3 III
Merck Index: 14,05136
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7e1dee5510dfc5aca5d5c8a3f82f993ab4d7216e&vid=7e1dee5510dfc5aca5d5c8a3f82f993ab4d7216e&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR