Malonamid ≥98.0% (by total nitrogen basis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

M0027-25G M0027-500G
TCIAM0027-25GEA 173 DKK
TCIAM0027-25G TCIAM0027-500G
Malonamid ≥98.0% (by total nitrogen basis)
Malonamid
Formel: C₃H₆N₂O₂
Kogepunkt: 461 °C (1013 hPa)
Smeltepunkt: 169…170 °C
Massefylde: 1,43 g/cm³ (20 °C)
MDL Number: MFCD00008034
CAS-nummer: 108-13-4
EINECS: 203-553-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=39cd475e6e3c433fe8d9889b1ea0d6f6fe920d17&vid=39cd475e6e3c433fe8d9889b1ea0d6f6fe920d17&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Melting point 169.0 to 175.0 deg-C
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR