(S)-(-)-1-Phenylethyl isocyanat ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: (S)-(-)-α-Phenethyl isocyanate, (S)-(-)-α-Methylbenzyl isocyanate, (S)-1-phenylethylisocyanate, (S)-(-)-1-Phenethyl isocyanate

I0335-1G I0335-5G
TCIAI0335-1GEA 279 DKK
TCIAI0335-1G TCIAI0335-5G
(S)-(-)-1-Phenylethyl isocyanat ≥98.0% (by GC)
(S)-(-)-1-Phenylethyl isocyanat
Formel: C₉H₉NO
Molekylvægt: 147.18 g/mol
Kogepunkt: 98°C
Flammepunkt: 65°C
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00002036
CAS-nummer: 14649-03-7
EINECS: 238-698-6
UN nummer: 2206
ADR: 6.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=284ef7530654509fc19340bf6c20d584c42aabb8&vid=284ef7530654509fc19340bf6c20d584c42aabb8&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %
Optical purity (GC) min. 98.0 ee%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR