Tetraethylammoniumfluorid, hydrat ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Tetrylammonium fluoride hydrate, Tetraethylammonium fluoride xhydrate

T0838-25G T0838-5G
TCIAT0838-25GEA 2340 DKK
TCIAT0838-25G TCIAT0838-5G
Tetraethylammoniumfluorid, hydrat ≥98.0% (by titrimetric analysis)
Tetraethylammoniumfluorid, hydrat
Formel: C₈H₂₀FN·xH₂O
Molekylvægt: 149.25 g/mol (anhy)
MDL Number: MFCD00149990
CAS-nummer: 98330-04-2
EINECS: 211-559-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=82076652f3608416a0007ecd9f2689dc8171dfb5&vid=82076652f3608416a0007ecd9f2689dc8171dfb5&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min 95.0% (calcd.on anh.substance)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR