1,3,5-Trioxan ≥99.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

T0505-500G T0505-25G
TCIAT0505-500GEA 386 DKK
TCIAT0505-500G TCIAT0505-25G
1,3,5-Trioxan ≥99.0% (by GC)
1,3,5-Trioxan
Formel: C₃H₆O₃
Kogepunkt: 115°C
Smeltepunkt: 60…62 °C
Massefylde: 1,17 g/cm³ (20 °C)
Flammepunkt: 38 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00006563
CAS-nummer: 110-88-3
EINECS: 203-812-5
UN nummer: 1325
ADR: 4.1,II
Merck Index: 14,09734
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fca59d9b4e7519512d283e0438bbfce2e2fbde6d&vid=fca59d9b4e7519512d283e0438bbfce2e2fbde6d&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White or Colorless powder to crystal
Purity (GC) min. 99.0 %
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR