Thiosemicarbazid ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

T0221-25G T0221-500G T0221-100G
TCIAT0221-25GEA 214 DKK
TCIAT0221-25G TCIAT0221-500G TCIAT0221-100G
Thiosemicarbazid ≥98.0% (by titrimetric analysis)
Thiosemicarbazid
Formel: CH₅N₃S
Smeltepunkt: 178…180 °C
MDL Number: MFCD00007620
CAS-nummer: 79-19-6
EINECS: 201-184-7
UN nummer: 2811
ADR: 6.1,II
Merck Index: 14,09361
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d243ebb31b8f2ca25fce07cd0d6736c977787e6b&vid=d243ebb31b8f2ca25fce07cd0d6736c977787e6b&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (Iodometric Titration) min. 98.0 %
Solubility in hot Water very faint turbidity
Solubility in hot Methanol very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR