Perylen ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
P0078-25G
P0078-5G
TCIAP0078-25GEA
2380
DKK
TCIAP0078-25G
TCIAP0078-5G
Perylen ≥98.0% (by GC)
Perylene
Formel:
C₂₀H₁₂ Kogepunkt: 443 °C (1013 hPa) Smeltepunkt: 273…278 °C Massefylde: 1,362 g/cm³ (20 °C) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00004142 CAS-nummer: 198-55-0 EINECS: 205-900-9 Merck Index: 14,07184 |
Specifikationer Testresultater
Appearance | Light yellow to Brown powder to crystal |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...