Acenaphthen ≥99.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
|
Synonymer:
1,2-Dihydroacenaphthylene
A0003-25G
A0003-500G
A0003-100G
TCIAA0003-25GEA
263
DKK
TCIAA0003-25G
TCIAA0003-500G
TCIAA0003-100G
Acenaphthen ≥99.0% (by GC)
Acenaphthen
Formel:
C₁₂H₁₀ Kogepunkt: 279 °C (1013 hPa) Smeltepunkt: 90…93 °C Massefylde: 1,024 g/cm³ (20 °C) Flammepunkt: (open cup) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00003807 CAS-nummer: 83-32-9 EINECS: 201-469-6 UN nummer: 3077 ADR: 9,III Merck Index: 14,00028 |
|
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 99.0 % |
Melting point | 93.0 to 96.0 deg-C |
Solubility in hot Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...