Tris(4-methylphenyl)phosphin ≥96.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Tri-p-tolylphosphine, Tris(p-tolyl)phosphine

T0862-25G T0862-5G
TCIAT0862-25GEA 920 DKK
TCIAT0862-25G TCIAT0862-5G
Tris(4-methylphenyl)phosphin ≥96.0% (by GC)
Tris(4-methylphenyl)phosphin
Formel: C₂₁H₂₁P
Molekylvægt: 304.37 g/mol
Kogepunkt: 400
Smeltepunkt: 144
MDL Number: MFCD00008542
CAS-nummer: 1038-95-5
EINECS: 213-863-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9f7f7cc60dad25dfe34104ade91e6a12a54e71fd&vid=9f7f7cc60dad25dfe34104ade91e6a12a54e71fd&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 96.0 %
Melting point 147.0 to 150.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR