Acetylthiocholiniodid ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

A0116-25G A0116-1G A0116-5G
TCIAA0116-25GEA 2390 DKK
TCIAA0116-25G TCIAA0116-1G TCIAA0116-5G
Acetylthiocholiniodid ≥98.0% (by titrimetric analysis)
Acetylthiocholiniodid
Formel: H₃CC(S)O(CH₂)₂N(CH₃)₃I
Smeltepunkt: 204 °C
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00011819
CAS-nummer: 1866-15-5
EINECS: 217-474-1
UN nummer: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=87d3f4d84d5d3b8bbe65be5d76730894b6566ecc&vid=87d3f4d84d5d3b8bbe65be5d76730894b6566ecc&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR