Trioctylphosphin ≥85.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

T0503-25ML T0503-500ML T0503-100ML
TCIAT0503-25MLEA 255 DKK
TCIAT0503-25ML TCIAT0503-500ML TCIAT0503-100ML
Trioctylphosphin ≥85.0% (by GC)
Trioctylphosphine
Formel: C₂₄H₅₁P
Kogepunkt: 234°C
Smeltepunkt: 48 °C
Massefylde: 0,831 g/cm³ (20 °C)
Flammepunkt: 147°C
MDL Number: MFCD00015298
CAS-nummer: 4731-53-7
EINECS: 225-234-2
UN nummer: 1760
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=694fafe0c96926a0eb4940cb209d6ac7e10e440f&vid=694fafe0c96926a0eb4940cb209d6ac7e10e440f&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 85.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR